Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012056.2 | NP_036186.2 | 2, 3 | 517 | ||
GenBank mRNA | AF090334.1 | - | 2, 3 | 443 | ||
AF279263.1 | - | 2, 3 | 495 | |||
AK184448.1 | - | 2, 3 | 188 | |||
AK198225.1 | - | 2, 3 | 187 | |||
AK198658.1 | - | 2, 3 | 187 | |||
AK204047.1 | - | 2, 3 | 187 | |||
AK206852.1 | - | 1, 2 | 68 | |||
AK208794.1 | - | 2, 3 | 184 | |||
AK209120.1 | - | 2, 3 | 187 | |||
AK209926.1 | - | 1, 2 | 137 | |||
AK214409.1 | - | 2, 3 | 187 | |||
AK216520.1 | - | 2, 3 | 187 | |||
BC026133.1 | - | 2, 3 | 457 | 4018235 | ||
BC043129.1 | - | 2, 3 | 517 | 6810029 |