Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082414.2 | NP_001075883.2 | 7 | 1579 | ||
GenBank mRNA | BC028254.1 | - | 7 | 1127 | ||
BC031117.1 | - | 7 | 690 | 5064197 | ||
D89677.2 | - | 13 | 2065 |