Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013463.2 | NP_038491.2 | 3 | 464 | ||
GenBank mRNA | AK040081.1 | - | 3 | 396 | ||
AK054547.1 | - | 3 | 416 | |||
AK145011.1 | - | 3 | 406 | |||
AK145148.1 | - | 464 | ||||
AK164231.1 | - | 3 | 397 | |||
AK166204.1 | - | 3 | 397 | |||
AK166245.1 | - | 3 | 413 | |||
BC009021.1 | - | 3 | 417 | 3708950 | ||
U34071.1 | - | 3 | 414 | |||
U50716.1 | - | 3 | 402 |