Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281845.1 | NP_001268774.1 | 3, 4 | 292 | ||
NM_013473.4 | NP_038501.2 | 3, 4 | 292 | |||
GenBank mRNA | AJ002390.1 | - | 3, 4 | 268 | ||
AK009085.1 | - | 3, 4 | 254 | |||
AK075870.1 | - | 3, 4 | 226 | |||
BC013271.1 | - | 3, 4 | 257 | 4013266 | ||
BC030407.1 | - | 3, 4 | 305 | 5322310 |