Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK133647.1 | - | 947 | |||
M37890.1 | - | 1 | 885 | |||
S56585.1 | - | 1 | 964 | |||
X53779.1 | - | 1 | 917 | |||
X59411.1 | - | 1 | 917 | |||
X59592.1 | - | 1 | 1005 |