Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2l10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013479.2 | NP_038507.1 | 1 | 463 | ||
GenBank mRNA | AF067660.1 | - | 1 | 460 | ||
AF102501.1 | - | 1 | 434 | |||
AK136172.1 | - | 1 | 464 | |||
AK145385.1 | - | 1 | 331 | |||
AK162127.1 | - | 1 | 331 | |||
BC052690.1 | - | 1 | 462 | 30052580 |