Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012001.2 | NP_036131.1 | 8, 9 | 1178 | ||
GenBank mRNA | AF071314.1 | - | 8, 9 | 1047 | ||
AK003752.1 | - | 8, 9 | 1045 | |||
AK009955.1 | - | 8, 9 | 1050 | |||
AK167833.1 | - | 8, 9 | 1064 | |||
BC020461.1 | - | 4, 5 | 1600 | |||
BC116800.1 | - | 8, 9 | 1032 | 40126119 | ||
BC116826.1 | - | 8, 9 | 1032 | 40126145 |