Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048208.1 | NP_001041673.1 | 6 | 982 | ||
NM_012005.3 | NP_036135.3 | 6 | 982 | |||
GenBank mRNA | AB019028.1 | - | 6 | 980 | ||
AK163306.1 | - | 7 | 744 | |||
BC065072.1 | - | 6 | 1082 | 6830820 |