Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012017.2 | NP_036147.1 | 7 | 3328 | ||
GenBank mRNA | AF083339.1 | - | 7 | 3339 | ||
AK028427.1 | - | 1 | 2486 | |||
AK033852.1 | - | 1 | 1657 | |||
AK084710.1 | - | 1 | 1803 | |||
BC055075.1 | - | 7 | 3322 | 30056643 |