Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTK2 SIRNA Assays ›
Gene Symbol:
PTK2
Gene Name:
protein tyrosine kinase 2
Gene Aliases:
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location:
Chr.8: 140658382 - 141001313 on Build GRCh38
UniGene ID:
Hs.395482
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
709
Purification
HPLC
Standard
In-Vivo Ready
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTK2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199649.1
NP_001186578.1
3
488
NM_001316342.1
NP_001303271.1
2
399
NM_005607.4
NP_005598.3
3
477
NM_153831.3
NP_722560.1
3
488
XM_005251003.2
XP_005251060.1
2
529
XM_005251012.2
XP_005251069.1
2
371
XM_005251013.2
XP_005251070.1
2
357
XM_006716608.2
XP_006716671.1
3
460
XM_006716609.2
XP_006716672.1
3
460
XM_006716610.2
XP_006716673.1
3
452
XM_011517194.2
XP_011515496.1
3
429
XM_017013651.1
XP_016869140.1
1
210
XM_017013652.1
XP_016869141.1
1
210
XM_017013653.1
XP_016869142.1
2
529
XM_017013654.1
XP_016869143.1
2
529
XM_017013655.1
XP_016869144.1
2
529
XM_017013656.1
XP_016869145.1
2
529
XM_017013657.1
XP_016869146.1
2
529
XM_017013658.1
XP_016869147.1
3
353
XM_017013659.1
XP_016869148.1
1
210
XM_017013660.1
XP_016869149.1
3
353
XM_017013661.1
XP_016869150.1
2
371
XM_017013662.1
XP_016869151.1
3
353
XM_017013663.1
XP_016869152.1
2
284
XM_017013664.1
XP_016869153.1
3
460
XM_017013665.1
XP_016869154.1
2
284
XM_017013666.1
XP_016869155.1
2
529
XM_017013667.1
XP_016869156.1
3
353
XM_017013668.1
XP_016869157.1
2
371
XM_017013669.1
XP_016869158.1
3
353
XM_017013670.1
XP_016869159.1
3
353
XM_017013671.1
XP_016869160.1
3
353
XM_017013672.1
XP_016869161.1
2
284
XM_017013673.1
XP_016869162.1
2
284
XM_017013674.1
XP_016869163.1
3
460
XM_017013675.1
XP_016869164.1
2
284
XM_017013676.1
XP_016869165.1
1
210
XM_017013677.1
XP_016869166.1
3
353
XM_017013678.1
XP_016869167.1
2
284
XM_017013679.1
XP_016869168.1
2
284
XM_017013680.1
XP_016869169.1
1
73
XM_017013681.1
XP_016869170.1
3
460
XM_017013682.1
XP_016869171.1
2
357
XM_017013683.1
XP_016869172.1
3
460
XM_017013684.1
XP_016869173.1
3
460
XM_017013685.1
XP_016869174.1
3
404
XM_017013686.1
XP_016869175.1
2
357
XM_017013687.1
XP_016869176.1
3
460
XM_017013688.1
XP_016869177.1
3
460
XM_017013689.1
XP_016869178.1
2
325
XM_017013690.1
XP_016869179.1
2
299
GenBank mRNA
AB209071.1
-
2
305
AK304356.1
-
3
333
AL832961.1
-
3
360
BC035404.2
-
1
209
4999680
BY800117.1
-
2
465
DA236314.1
-
3
460
DA994616.1
-
2
258
KF437463.1
-
1
181
L13616.1
-
3
410
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001130409, NM_007982, AK079821, AB011499, AB030035, and others.
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
microtubule cytoskeleton organization
angiogenesis
vasculogenesis
neuron migration
placenta development
regulation of protein phosphorylation
positive regulation of protein phosphorylation
heart morphogenesis
cellular component disassembly involved in execution phase of apoptosis
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
integrin-mediated signaling pathway
axon guidance
positive regulation of cell proliferation
regulation of cell shape
embryo development
regulation of endothelial cell migration
regulation of epithelial cell migration
positive regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
central nervous system neuron axonogenesis
negative regulation of cell-cell adhesion
establishment of cell polarity
extracellular matrix organization
positive regulation of cell migration
regulation of cell adhesion mediated by integrin
netrin-activated signaling pathway
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
establishment of nucleus localization
regulation of cell proliferation
negative regulation of apoptotic process
regulation of GTPase activity
endothelial cell migration
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
innate immune response
regulation of osteoblast differentiation
positive regulation of protein kinase activity
negative regulation of organ growth
protein autophosphorylation
vascular endothelial growth factor receptor signaling pathway
ephrin receptor signaling pathway
cell motility
negative regulation of axonogenesis
regulation of cytoskeleton organization
regulation of focal adhesion assembly
positive regulation of protein kinase B signaling
negative regulation of synapse assembly
growth hormone receptor signaling pathway
regulation of substrate adhesion-dependent cell spreading
positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of anoikis
actin binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
signal transducer activity
Ras guanyl-nucleotide exchange factor activity
receptor binding
protein binding
ATP binding
JUN kinase binding
protein kinase binding
SH2 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline