Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tinag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012033.3 | NP_036163.3 | 7 | 1019 | ||
GenBank mRNA | AF153366.1 | - | 7 | 961 | ||
AK008374.1 | - | 7 | 991 | |||
BC010745.1 | - | 7 | 964 | 4239192 |