Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002013.3 | NP_002004.1 | 1 | 171 | ||
XM_011536565.2 | XP_011534867.1 | 1 | 102 | |||
XM_017021092.1 | XP_016876581.1 | 1 | 120 | |||
XM_017021093.1 | XP_016876582.1 | 1 | 102 | |||
GenBank mRNA | AK222993.1 | - | 1 | 120 | ||
AK311915.1 | - | 1 | 109 | |||
BC016288.1 | - | 2 | 509 | 3919642 | ||
BC020809.1 | - | 1 | 109 | 4292678 | ||
BI598048.1 | - | 1 | 176 | |||
BT006904.1 | - | 1 | 98 | |||
M96256.1 | - | 1 | 121 |