Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013468.3 | NP_038496.2 | 3 | 395 | ||
GenBank mRNA | AF041847.1 | - | 3 | 346 | ||
AF041849.1 | - | 1 | 27 | |||
AK009655.1 | - | 3 | 388 | |||
AK009959.1 | - | 3 | 394 | |||
AK145944.1 | - | 3 | 436 | |||
AK146471.1 | - | 3 | 392 | |||
AK146627.1 | - | 3 | 394 | |||
AK146940.1 | - | 3 | 392 | |||
BC037138.1 | - | 3 | 395 | 4953125 |