Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013470.2 | NP_038498.2 | 10 | 913 | ||
GenBank mRNA | AJ001633.1 | - | 10 | 896 | ||
AK013850.1 | - | 8 | 666 | |||
AK090055.1 | - | 10 | 931 | |||
AK150922.1 | - | 10 | 914 | |||
AK150951.1 | - | 10 | 881 | |||
AK151771.1 | - | 10 | 873 | |||
AK152837.1 | - | 10 | 840 | |||
AK152863.1 | - | 10 | 901 | |||
AK166389.1 | - | 10 | 885 | |||
AK168396.1 | - | 10 | 809 | |||
AK168507.1 | - | 10 | 841 | |||
AK169113.1 | - | 10 | 789 | |||
BC090634.1 | - | 10 | 907 | 30942562 | ||
BC141167.1 | - | 10 | 867 | 9055779 | ||
CT010280.1 | - | 8 | 696 |