Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bop1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013481.1 | NP_038509.1 | 16 | 2416 | ||
GenBank mRNA | AK079627.1 | - | 16 | 2396 | ||
AK129058.1 | - | 16 | 2389 | |||
AK144095.1 | - | 16 | 2428 | |||
AK167110.1 | - | 16 | 2399 | |||
AK167878.1 | - | 16 | 2404 | |||
AK168127.1 | - | 16 | 2397 | |||
AK187459.1 | - | 1 | 203 | |||
AK191694.1 | - | 1 | 203 | |||
AK198058.1 | - | 1 | 203 | |||
AK199109.1 | - | 1 | 202 | |||
BC012693.1 | - | 16 | 2390 | 4193123 | ||
U77415.1 | - | 16 | 2416 |