Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198859.1 | NP_001185788.1 | 2, 3 | 374 | ||
NM_001198860.1 | NP_001185789.1 | 3, 4 | 567 | |||
NM_001198861.1 | NP_001185790.1 | 3, 4 | 567 | |||
NM_013502.3 | NP_038530.1 | 2, 3 | 374 | |||
GenBank mRNA | AB033122.1 | - | 2, 3 | 339 | ||
AJ010483.1 | - | 2, 3 | 287 | |||
AK133816.1 | - | 3, 4 | 538 | |||
AK149906.1 | - | 3, 4 | 545 | |||
AK160658.1 | - | 3, 4 | 557 | |||
AK165276.1 | - | 3, 4 | 522 | |||
AK170133.1 | - | 1, 2 | 81 | |||
AK178530.1 | - | 1, 2 | 83 | |||
AK179724.1 | - | 1, 2 | 110 | |||
AK214870.1 | - | 1, 2 | 83 | |||
AK217104.1 | - | 1, 2 | 101 | |||
AK218288.1 | - | 1, 2 | 86 | |||
AK219771.1 | - | 1, 2 | 83 | |||
BC013702.1 | - | 2, 3 | 287 | 3155102 | ||
BC015071.1 | - | 2, 3 | 343 | 4192052 | ||
BC042425.1 | - | 3, 4 | 548 | 5005681 |