Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013505.3 | NP_038533.1 | 3 | 581 | ||
GenBank mRNA | AK028604.1 | - | 3 | 586 | ||
AK028636.1 | - | 3 | 582 | |||
AK030333.1 | - | 3 | 582 | |||
AK048547.1 | - | 3 | 582 | |||
AK172168.1 | - | 3 | 416 | |||
BC057867.1 | - | 3 | 361 | 5327748 | ||
L33779.1 | - | 3 | 380 |