Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199265.1 | NP_001186194.1 | 4 | 902 | ||
NM_013511.3 | NP_038539.2 | 4 | 793 | |||
GenBank mRNA | AB370258.1 | - | 3 | 704 | ||
AB370259.1 | - | 3 | 704 | |||
AB370260.1 | - | 3 | 704 | |||
AB370263.1 | - | 3 | 704 | |||
AF044312.1 | - | 3 | 704 | |||
AJ539142.1 | - | 3 | 704 | |||
AJ539143.1 | - | 3 | 704 | |||
AJ539144.1 | - | 3 | 704 | |||
AJ542537.1 | - | 3 | 704 | |||
AK043148.1 | - | 3 | 681 | |||
AK052475.1 | - | 4 | 793 | |||
AK144088.1 | - | 4 | 903 | |||
AK167434.1 | - | 4 | 819 | |||
AK170275.1 | - | 4 | 845 | |||
BC023220.1 | - | 4 | 789 | |||
BC023768.1 | - | 4 | 815 | |||
BC026708.1 | - | 4 | 774 | |||
BC026714.1 | - | 3 | 386 | |||
BC029938.1 | - | 4 | 931 | |||
KF534728.1 | - | 3 | 704 |