Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013513.2 | NP_038541.1 | 3 | 455 | ||
GenBank mRNA | AK133896.1 | - | 3 | 469 | ||
AK137484.1 | - | 3 | 536 | |||
AK143841.1 | - | 3 | 435 | |||
AK158219.1 | - | 3 | 373 | |||
AK158247.1 | - | 3 | 419 | |||
AK158370.1 | - | 3 | 382 | |||
AK165288.1 | - | 3 | 382 | |||
AK172410.1 | - | 3 | 419 | |||
BC053739.1 | - | 3 | 426 | |||
BC066194.1 | - | 3 | 455 | 30429971 | ||
BC066806.1 | - | 3 | 455 | 30429961 | ||
L35933.1 | - | 3 | 434 | |||
U03487.1 | - | 3 | 385 | |||
U04055.1 | - | 3 | 455 |