Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276328.1 | NP_001263257.1 | 6, 7 | 604 | ||
NM_001276330.1 | NP_001263259.1 | 6, 7 | 616 | |||
NM_013516.2 | NP_038544.1 | 6, 7 | 715 | |||
GenBank mRNA | AK144021.1 | - | 6, 7 | 605 | ||
BC139323.1 | - | 6, 7 | 639 | 8862345 | ||
BC139324.1 | - | 6, 7 | 639 | 8862346 | ||
CN663232.1 | - | 6, 7 | 556 | |||
J05019.1 | - | 6, 7 | 639 |