Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013520.3 | NP_038548.3 | 5 | 370 | ||
GenBank mRNA | AK020105.1 | - | 5 | 370 | ||
BC019801.1 | - | 5 | 355 | 5132319 | ||
CK622267.1 | - | 5 | 340 | |||
EU274583.1 | - | 4 | 282 | |||
L23636.1 | - | 4 | 313 | |||
S76459.1 | - | 4 | 288 | |||
U04807.1 | - | 4 | 537 |