Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 4 | 986 | ||
NM_001143677.1 | NP_001137149.1 | 2 | 237 | |||
NM_001143678.1 | NP_001137150.1 | 2 | 416 | |||
NM_001291995.1 | NP_001278924.1 | 2 | 192 | |||
NM_005627.3 | NP_005618.2 | 2 | 192 | |||
XM_011536071.1 | XP_011534373.1 | 5 | 768 | |||
GenBank mRNA | AF153609.1 | - | 2 | 145 | ||
AK055077.1 | - | 4 | 986 | |||
AK098509.1 | - | 2 | 160 | |||
AK293383.1 | - | 2 | 226 | |||
AK295422.1 | - | 2 | 160 | |||
AK298688.1 | - | 2 | 475 | |||
BC001263.1 | - | 2 | 145 | 3459056 | ||
BX649005.1 | - | 2 | 163 | |||
DA309338.1 | - | 2 | 270 | |||
DC428953.1 | - | 2 | 475 | |||
EU518415.1 | - | 4 | 388 | |||
FM205707.1 | - | 2 | 180 | |||
FM205708.1 | - | 2 | 215 | |||
FM205709.1 | - | 4 | 835 | |||
FM205710.1 | - | 1 | 304 | |||
JQ836654.1 | - | 2 | 145 | |||
Y10032.1 | - | 2 | 145 |