Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hebp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013546.2 | NP_038574.2 | 1, 2 | 204 | ||
GenBank mRNA | AA022130.1 | - | 192 | |||
AB013095.1 | - | 1, 2 | 143 | |||
AF117613.1 | - | 1, 2 | 192 | |||
AK008919.1 | - | 1, 2 | 125 | |||
BC012654.1 | - | 1, 2 | 128 | 4216585 | ||
BY242687.1 | - | 1, 2 | 205 | |||
CT010262.1 | - | 1, 2 | 86 |