Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161824.1 | NP_001155296.1 | 4 | 715 | ||
NM_013604.2 | NP_038632.2 | 4 | 718 | |||
GenBank mRNA | AK086135.1 | - | 4 | 311 | ||
AK159316.1 | - | 3 | 405 | |||
AK163982.1 | - | 4 | 286 | |||
AK165547.1 | - | 4 | 293 | |||
AK165923.1 | - | 4 | 291 | |||
AX775987.1 | - | 4 | 290 | |||
BB662550.1 | - | 4 | 310 | |||
BC023071.1 | - | 4 | 289 | 5355724 | ||
L36962.1 | - | 4 | 288 |