Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Muc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013605.2 | NP_038633.1 | 2 | 153 | ||
GenBank mRNA | BC005441.1 | - | 2 | 124 | 3601782 | |
M84683.1 | - | 2 | 103 |