Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crem | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110850.2 | NP_001104320.1 | 5 | 471 | ||
NM_001110853.2 | NP_001104323.1 | 7 | 734 | |||
NM_001110854.1 | NP_001104324.2 | 3 | 113 | |||
NM_001110855.1 | NP_001104325.2 | 2 | 77 | |||
NM_001110856.2 | NP_001104326.1 | 7 | 654 | |||
NM_001110857.2 | NP_001104327.1 | 6 | 507 | |||
NM_001110858.2 | NP_001104328.1 | 5 | 471 | |||
NM_001110859.2 | NP_001104329.1 | 8 | 843 | |||
NM_001271503.1 | NP_001258432.1 | 6 | 618 | |||
NM_001271504.1 | NP_001258433.1 | 6 | 618 | |||
NM_001271505.1 | NP_001258434.1 | 7 | 696 | |||
NM_001271506.1 | NP_001258435.1 | 7 | 807 | |||
NM_013498.3 | NP_038526.2 | 6 | 507 | |||
GenBank mRNA | AJ292222.1 | - | 2 | 42 | ||
AJ311667.1 | - | 3 | 78 | |||
AK076797.1 | - | 7 | 657 | |||
AY738715.1 | - | 5 | 534 | |||
AY738716.1 | - | 4 | 387 | |||
AY738717.1 | - | 5 | 534 | |||
AY738718.1 | - | 6 | 570 | |||
AY738719.1 | - | 6 | 612 | |||
AY738720.1 | - | 6 | 630 | |||
AY738721.1 | - | 6 | 723 | |||
BE689077.1 | - | 2 | 77 | |||
M60285.1 | - | 6 | 435 | |||
S64590.1 | - | 6 | 552 | |||
S67786.1 | - | 2 | 64 |