Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198859.1 | NP_001185788.1 | 2, 3 | 389 | ||
NM_001198860.1 | NP_001185789.1 | 3, 4 | 582 | |||
NM_001198861.1 | NP_001185790.1 | 3, 4 | 582 | |||
NM_013502.3 | NP_038530.1 | 2, 3 | 389 | |||
GenBank mRNA | AB033122.1 | - | 2, 3 | 354 | ||
AJ010483.1 | - | 2, 3 | 302 | |||
AK133816.1 | - | 3, 4 | 553 | |||
AK149906.1 | - | 3, 4 | 560 | |||
AK160658.1 | - | 3, 4 | 572 | |||
AK165276.1 | - | 3, 4 | 537 | |||
AK170133.1 | - | 1, 2 | 96 | |||
AK178530.1 | - | 1, 2 | 98 | |||
AK179724.1 | - | 1, 2 | 125 | |||
AK206815.1 | - | 1, 2 | 99 | |||
AK214870.1 | - | 1, 2 | 98 | |||
AK217104.1 | - | 1, 2 | 116 | |||
AK218288.1 | - | 1, 2 | 101 | |||
AK219771.1 | - | 1, 2 | 98 | |||
BC013702.1 | - | 2, 3 | 302 | 3155102 | ||
BC015071.1 | - | 2, 3 | 358 | 4192052 | ||
BC042425.1 | - | 3, 4 | 563 | 5005681 |
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK179015.1 | - | 98 |