Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013505.3 | NP_038533.1 | 3, 4 | 615 | ||
GenBank mRNA | AK028636.1 | - | 3, 4 | 616 | ||
AK030333.1 | - | 3, 4 | 616 | |||
AK048547.1 | - | 3, 4 | 616 | |||
AK172168.1 | - | 3, 4 | 450 | |||
BC057867.1 | - | 3, 4 | 395 | 5327748 | ||
L33779.1 | - | 3, 4 | 414 |