Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276328.1 | NP_001263257.1 | 7 | 718 | ||
NM_001276329.1 | NP_001263258.1 | 6 | 499 | |||
NM_001276330.1 | NP_001263259.1 | 7 | 730 | |||
NM_013516.2 | NP_038544.1 | 7 | 829 | |||
GenBank mRNA | AK144021.1 | - | 7 | 719 | ||
AK161121.1 | - | 6 | 465 | |||
J05019.1 | - | 7 | 753 | |||
U90218.1 | - | 2 | 191 |