Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013523.3 | NP_038551.3 | 3 | 323 | ||
GenBank mRNA | AF095642.1 | - | 3 | 260 | ||
AK016635.1 | - | 3 | 323 | |||
BC137990.1 | - | 3 | 320 | 8861009 | ||
BC137991.1 | - | 3 | 320 | 8861010 |