Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013536.2 | NP_038564.1 | 2 | 346 | ||
GenBank mRNA | AK081757.1 | - | 2 | 324 | ||
BC002004.1 | - | 2 | 266 | 3487480 | ||
CB948801.1 | - | 2 | 349 |