Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013538.5 | NP_038566.1 | 4 | 575 | ||
GenBank mRNA | AK008606.1 | - | 4 | 528 | ||
AK010409.1 | - | 4 | 516 | |||
AK011313.1 | - | 4 | 548 | |||
AK028159.1 | - | 4 | 517 | |||
AK145537.1 | - | 4 | 517 | |||
AK169792.1 | - | 4 | 523 | |||
BC002006.1 | - | 4 | 529 | 3592108 | ||
BC027172.1 | - | 3 | 1153 |