Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143676.1 | NP_001137148.1 | 14 | 2379 | ||
NM_001143677.1 | NP_001137149.1 | 12 | 1630 | |||
NM_001143678.1 | NP_001137150.1 | 12 | 1809 | |||
NM_001291995.1 | NP_001278924.1 | 11 | 1453 | |||
NM_005627.3 | NP_005618.2 | 12 | 1585 | |||
XM_011536071.1 | XP_011534373.1 | 15 | 2161 | |||
GenBank mRNA | AF153609.1 | - | 12 | 1538 | ||
AK055077.1 | - | 14 | 2379 | |||
AK098509.1 | - | 12 | 1553 | |||
BC001263.1 | - | 12 | 1538 | 3459056 | ||
BX649005.1 | - | 2 | 4655 | |||
FM205707.1 | - | 12 | 1573 | |||
FM205708.1 | - | 12 | 1608 | |||
FM205709.1 | - | 14 | 2228 | |||
FM205710.1 | - | 11 | 1697 | |||
Y10032.1 | - | 12 | 1537 |