Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013556.2 | NP_038584.2 | 2, 3 | 288 | ||
GenBank mRNA | AK002286.1 | - | 2, 3 | 233 | ||
AK082827.1 | - | 2, 3 | 178 | |||
AK088114.1 | - | 2, 3 | 178 | |||
AK146626.1 | - | 2, 3 | 234 | |||
BC004686.1 | - | 2, 3 | 203 | 3500885 | ||
BC083145.1 | - | 2, 3 | 174 | 30445781 | ||
BY285280.1 | - | 2, 3 | 289 | |||
J00423.1 | - | 2, 3 | 229 |