Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000220.4 | NP_000211.1 | 2 | 2056 | ||
NM_153764.2 | NP_722448.1 | 2 | 2104 | |||
NM_153765.2 | NP_722449.3 | 3 | 2237 | |||
NM_153766.2 | NP_722450.1 | 3 | 2274 | |||
NM_153767.3 | NP_722451.1 | 4 | 2407 | |||
GenBank mRNA | BC063109.1 | - | 4 | 2408 | ||
U03884.1 | - | 3 | 2184 |