Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198859.1 | NP_001185788.1 | 9 | 1807 | ||
NM_001198860.1 | NP_001185789.1 | 10 | 2000 | |||
NM_001198861.1 | NP_001185790.1 | 10 | 2003 | |||
NM_013502.3 | NP_038530.1 | 9 | 1810 | |||
GenBank mRNA | AB033122.1 | - | 9 | 1775 | ||
AJ010483.1 | - | 9 | 1720 | |||
AK133816.1 | - | 10 | 1974 | |||
AK149906.1 | - | 7 | 3417 | |||
AK160658.1 | - | 10 | 1990 | |||
AK170133.1 | - | 8 | 1517 | |||
AK171650.1 | - | 8 | 1643 | |||
AK216177.1 | - | 1 | 155 | |||
BC013702.1 | - | 9 | 1723 | 3155102 | ||
BC015071.1 | - | 9 | 1779 | 4192052 | ||
BC042425.1 | - | 10 | 1984 | 5005681 |