Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb4.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013513.2 | NP_038541.1 | 13 | 2770 | ||
GenBank mRNA | AK080230.1 | - | 4 | 1056 | ||
AK089080.1 | - | 7 | 1818 | |||
AK133896.1 | - | 13 | 2785 | |||
AK143841.1 | - | 13 | 2750 | |||
AK158219.1 | - | 13 | 2688 | |||
AK158247.1 | - | 13 | 2734 | |||
AK165288.1 | - | 13 | 2697 | |||
AK172410.1 | - | 13 | 2734 | |||
BC053739.1 | - | 13 | 2742 | |||
BC066194.1 | - | 13 | 2770 | 30429971 | ||
BC066806.1 | - | 13 | 2770 | 30429961 | ||
L35933.1 | - | 13 | 2749 | |||
U04055.1 | - | 13 | 2769 |