Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013531.4 | NP_038559.2 | 10 | 2267 | ||
GenBank mRNA | AK036816.1 | - | 10 | 2269 | ||
AK098139.1 | - | 10 | 2201 | |||
AK152227.1 | - | 10 | 2258 | |||
AK155309.1 | - | 10 | 2333 | |||
AK158965.1 | - | 11 | 2496 | |||
AK168302.1 | - | 10 | 2227 | |||
AK169752.1 | - | 9 | 2349 | |||
AK211383.1 | - | 1 | 375 |