Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013538.5 | NP_038566.1 | 6 | 902 | ||
GenBank mRNA | AK008606.1 | - | 6 | 855 | ||
AK010409.1 | - | 6 | 843 | |||
AK011313.1 | - | 6 | 875 | |||
AK028159.1 | - | 6 | 844 | |||
AK145537.1 | - | 6 | 844 | |||
AK169792.1 | - | 6 | 850 | |||
BC002006.1 | - | 6 | 856 | 3592108 | ||
BC027172.1 | - | 5 | 1480 |