Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013557.2 | NP_038585.2 | 903 | |||
GenBank mRNA | AK156492.1 | - | 862 | |||
AK173160.1 | - | 853 | ||||
AK215017.1 | - | 4 | 252 | |||
AY033898.1 | - | 888 | ||||
BC028923.1 | - | 875 | 4164417 | |||
BC111035.1 | - | 854 | 5357121 |