Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013581.3 | NP_038609.3 | 9, 10 | 2401 | ||
GenBank mRNA | AF109377.2 | - | 9, 10 | 2384 | ||
AK031510.1 | - | 8, 9 | 1819 | |||
AK034636.2 | - | 9, 10 | 2406 | |||
AK152464.1 | - | 9, 10 | 2392 | |||
AK158507.1 | - | 9, 10 | 2763 | |||
AK159497.1 | - | 9, 10 | 2403 | |||
AK162971.1 | - | 9, 10 | 2402 | |||
AK171574.1 | - | 9, 10 | 2395 | |||
AK177622.1 | - | 1, 2 | 170 | |||
AK187486.1 | - | 1, 2 | 170 | |||
AK189116.1 | - | 1, 2 | 170 | |||
AK217864.1 | - | 1, 2 | 170 | |||
AK220387.1 | - | 8, 9 | 2845 | |||
BC049541.1 | - | 3, 4 | 851 | |||
BC063056.1 | - | 9, 10 | 2383 | 5720460 |