Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr4a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139509.1 | NP_001132981.1 | 6 | 1612 | ||
NM_013613.2 | NP_038641.1 | 6 | 1758 | |||
GenBank mRNA | AB014889.1 | - | 6 | 1351 | ||
AK087495.1 | - | 7 | 1636 | |||
AK158607.1 | - | 6 | 1759 | |||
BC137715.1 | - | 6 | 2070 | 8860734 | ||
BC144854.1 | - | 6 | 2286 | |||
S53744.1 | - | 6 | 1649 |