Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pafah1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013625.4 | NP_038653.1 | 3 | 650 | ||
GenBank mRNA | AK144908.1 | - | 3 | 639 | ||
AK147207.1 | - | 3 | 588 | |||
AK147229.1 | - | 3 | 601 | |||
AK147237.1 | - | 3 | 642 | |||
AK147309.1 | - | 3 | 585 | |||
AK147507.1 | - | 3 | 924 | |||
AK147515.1 | - | 3 | 637 | |||
AK147533.1 | - | 3 | 617 | |||
AK147793.1 | - | 3 | 312 | |||
AK160764.1 | - | 3 | 867 | |||
AK164008.1 | - | 3 | 321 | |||
AK167448.1 | - | 3 | 617 | |||
AK167824.1 | - | 3 | 642 | |||
AK168929.1 | - | 3 | 290 | |||
AK168960.1 | - | 3 | 605 | |||
AY189217.1 | - | 2 | 269 | |||
AY189218.1 | - | 2 | 269 | |||
BC014831.1 | - | 3 | 589 | 4507474 | ||
BC026141.1 | - | 3 | 614 | 4017963 | ||
BC046989.1 | - | 3 | 623 | |||
BC099462.1 | - | 3 | 589 | |||
BQ895724.1 | - | 3 | 650 | |||
L25108.1 | - | 3 | 550 | |||
U95116.1 | - | 3 | 945 |