Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048200.2 | NP_001041665.1 | 2 | 752 | ||
NM_001278162.1 | NP_001265091.1 | 2 | 544 | |||
NM_001278163.1 | NP_001265092.1 | 2 | 265 | |||
NM_005734.4 | NP_005725.3 | 2 | 752 | |||
XM_005252729.2 | XP_005252786.1 | 2 | 539 | |||
XM_017017076.1 | XP_016872565.1 | 1 | 186 | |||
XM_017017077.1 | XP_016872566.1 | 2 | 209 | |||
GenBank mRNA | AF004849.1 | - | 1 | 191 | ||
AF305239.1 | - | 2 | 215 | |||
AK293009.1 | - | 2 | 545 | |||
AK315277.1 | - | 2 | 265 | |||
BF332205.1 | - | 1 | 216 | |||
KU921433.1 | - | 1 | 184 | |||
Y09306.1 | - | 2 | 234 |