Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110780.1 | NP_001104250.1 | 2 | 531 | ||
NM_013680.4 | NP_038708.3 | 2 | 531 | |||
GenBank mRNA | BB646141.1 | - | 2 | 532 | ||
BC022954.1 | - | 2 | 509 | 5368098 |