Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013684.3 | NP_038712.3 | 3 | 364 | ||
GenBank mRNA | AK085037.1 | - | 3 | 365 | ||
BC012685.1 | - | 5 | 600 | 3498542 | ||
BC016476.1 | - | 3 | 342 | 3584341 | ||
BC050136.1 | - | 3 | 315 | 30020603 | ||
D01034.1 | - | 2 | 181 | |||
FJ770049.1 | - | 2 | 203 | |||
U63933.1 | - | 4 | 364 |