Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trhr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013696.2 | NP_038724.1 | 2 | 432 | ||
GenBank mRNA | BB653069.1 | - | 2 | 433 | ||
BC109005.1 | - | 2 | 317 | 40048176 | ||
BC109006.1 | - | 2 | 317 | 40048178 | ||
BC128269.1 | - | 2 | 317 | 40048433 | ||
M59811.1 | - | 2 | 341 | |||
M94384.1 | - | 2 | 341 |
Set Membership: |
G-protein coupled receptor |