Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000744.6 | NP_000735.1 | 6 | 3326 | ||
NM_001256573.1 | NP_001243502.1 | 6 | 3257 | |||
NR_046317.1 | 6 | 3351 | ||||
XM_011528524.1 | XP_011526826.1 | 5 | 2970 | |||
XM_017027624.1 | XP_016883113.1 | 7 | 3358 | |||
XM_017027625.1 | XP_016883114.1 | 7 | 3376 | |||
GenBank mRNA | U62433.1 | - | 7 | 2914 |