Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164274.1 | NP_001157746.1 | 2 | 288 | ||
NM_013720.2 | NP_038748.2 | 2 | 288 | |||
GenBank mRNA | AF205935.1 | - | 1 | 102 | ||
AK012050.1 | - | 2 | 251 | |||
AK032570.1 | - | 2 | 269 | |||
AK046252.1 | - | 2 | 289 | |||
AK133775.1 | - | 2 | 505 | |||
AK139919.1 | - | 2 | 240 | |||
AK145491.1 | - | 2 | 237 | |||
AK162302.1 | - | 2 | 285 | |||
AK172962.1 | - | 2 | 211 |