Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013626.3 | NP_038654.2 | 4, 5 | 725 | ||
GenBank mRNA | AK049423.1 | - | 3, 4 | 386 | ||
AK051649.1 | - | 4, 5 | 715 | |||
U79523.1 | - | 4, 5 | 610 |